SP Bel-Art Observation Fume Hood, Bel-Art Products, a part of SP
Supplier: Bel-Art
H50000-2002
74950-354EA 976.16 USD
74950-354
SP Bel-Art Observation Fume Hood, Bel-Art Products, a part of SP
Fume Hoods and Enclosures Chemical Fume Hoods
Lightweight and economical, this hood has clear polycarbonate windows which are stronger than acrylic, wipe clean with a damp cloth, and provide excellent visualization in a group setting.
  • Polyethylene plastic surface
  • Clear polycarbonate windows
  • Easy to clean
  • Lightweight and economical
  • Chemical-resistant hood surface
The front sash opens to three adjustable positions for air flow control. The hood’s polyethylene plastic surface is chemical resistant and easy to clean.
Información para pedidos: Stepped exhaust duct will accept either a 12 or 15 cm (4 or 6") diameter duct. An exhaust system capable of drawing 200 cfm (free air) is recommended.