MEP HyperCel® Hydrophobic Charge Induction Chromatography (HCIC) Resin, Pall® Life Sciences
Supplier: Cytiva
12035-028 12035-028 12035-010 12035-010
87004-854EA 670.52 USD
CA87004-852 87004-854 87004-852 CA87004-854
MEP HyperCel® Hydrophobic Charge Induction Chromatography (HCIC) Resin, Pall® Life Sciences
MEP (mercaptoethylpyridine) HyperCel® resin is a high-capacity, high-selectivity resin specially designed for the capture and purification of monoclonal and polyclonal antibodies. Resin supports efficient capture and purification of antibodies from a broad range of sources, such as animal sera, ascites fluids, and cell culture supernatant. A variety of cell culture formulations are readily accommodated, including protein-free, albumin-supplemented, and serum-supplemented media. In contrast to Protein A resins, IgG binding capacity on MEP HyperCel® resin is essentially independent of subclass or species. Weakly-binding variants (e.g. murine IgG, rat IgGs) are well retained. Dynamic binding capacity determined using 5 mg/mL human IgG in PBS; flow rate 60 cm/h.
  • For monoclonal and polyclonal antibodies
  • Supports efficient capture and purification
  • Accommodates a variety of cell culture formulations
MEP HyperCel® resin provides significant benefits at both laboratory and process scale. Features also include direct sample loading without any adjustment of pH or ionic strength, high purity achieved in a single step, high IgG capacity, and easy cleaning with sodium hydroxide.
This resin can also be an excellent alternative to traditional HIC resins for capture of hydrophobic molecules which are non-antibody proteins, or for impurity removal.
MEP HyperCel™ resin provides significant benefits at both laboratory and process scale. Features also include direct sample loading without any adjustment of pH or ionic strength, high purity achieved in a single step, high IgG capacity, and easy cleaning with sodium hydroxide.