Spiral Tube, Labconco
Supplier: Labconco
7027700
30182-417EA 2135 USD
30182-417
Spiral Tube, Labconco
Ducting Supplies
For temporary installations of laboratory hoods.
Neoprene-impregnated fiberglass reinforced with steel wire. Length: 3.1 m (10').
Información de suministro: Includes rigid duct connector and two clamps.