VWR® Critical Storage Undercounter Laboratory Refrigerator and Freezer units are designed to meet the demanding standards of clinical, life science, pharmaceutical, biotechnology, and industrial applications.
- Optimal performance in clinical, life science, pharma, biotechnology, and industrial applications
- Undercounter, freestanding, or stacking options
- Exceptional temperature control and uniformity
- Microprocessor control with user-programmable functions and features
- Built-in front-breather condenser
Units offer exceptional temperature control and uniformity via a door-mounted microprocessor controller and interior forced air circulation
A built-in front-breather condenser allows units to be placed under benches or countertops and requires no additional clearance from the top, back, or sides of the cabinet. These compact units can also be left free standing, placed side by side, or securely stacked to create a combination refrigerator/freezer. A keyed door ensures safe and secure storage within each unit. The refrigerator is equipped with adjustable wire shelves, the freezer is equipped with fixed evaporator shelves, and both units feature a right-hinged door swing and a rounded door profile and handle.
The easy-to-read controller features an upward angled LED display and keypad. Digital temperature input allows end user temperature flexibility. The controller lockout function prevents set point tampering. A preset alarm at ±3 °C from temperature set point monitors critical variances. A door-ajar alarm with alarm delay timer eliminates nuisance alarms, and remote alarm contacts allow for connection to a centralized alarm monitoring system.
Certificaciones: Both models meet JCAHO standards for controlling medication access.
Información accesorios: A stacking kit is available separately.
Información para pedidos: A 3 cm (13/16") access port is standard.
Precaución: Due to recent enactment in the State of California a new regulation effective January 1, 2020 that prohibits certain products manufactured after December 31, 2019 from being sold into California. The criteria of the regulation are based on gases used in the manufacture of the product which bans this product from California. As of January 1, 2020, Avantor will no longer offer these products for purchase in the entire State of California, effective January 1, 2020.