Freedom™ 2000 Series Reusable Half Mask Respirators, Honeywell Safety
Supplier: Honeywell Safety
Freedom™
14150098 14150091 14150097 14150087 14150087
56611-658CS 559.66 USD
56611-658 CA56611-658 CA56611-650 CA56611-662 CA56611-664
Freedom™ 2000 Series Reusable Half Mask Respirators, Honeywell Safety
Respirators Reusable Respirators Half Face Respirators
Disposable respirator with very low inhalation breathing resistance.
Anatomically-correct cradle strap, anti-slip headband locks and sliding suspension system allow easy sizing and quick donning and removal. Rolled edges and broad sealing flanges enhance comfort, assure a proper fit. Preassembled. Permanently affixed cartridges are color-coded for hazards.