SureGrip™ Alligator Clips, Model AC285, Fluke®
Supplier: Fluke
SureGrip™
1610159
26984-312EA 72.06 USD
26984-312
SureGrip™ Alligator Clips, Model AC285, Fluke®
Electrical Sockets
Sold as a pair containing one red and one black clip.
Multi-purpose tooth pattern grips anything from fine gauge wire to a 1.9 cm (3/4") bolt. Nickel-plated steel jaws. CAT III rated for 1000 V; CAT IV rated for 600 V, 10 A.