Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
/
Puerto Rico
Products
Services
Digital Solutions
Applications
Support
About
$0.00
Order Entry
Home
Standards
Organic Standards
rac Ketoprofen, TRC, LGC Standards
rac Ketoprofen, TRC, LGC Standards
rac Ketoprofen, TRC, LGC Standards
77694-621
:
LGC Standards
New Product
:
rac Ketoprofen, TRC, LGC Standards
77694-621
:
LGC Standards
:
TRC-K200800-1G
New Product
:
$88.70
Each(1/Itm/Bx)
-
+
CAS No.:
22071-15-4
Description:
rac Ketoprofen
Size:
1 g
Formula:
C₁₆H₁₄O₃
Molecular Weight:
254.29 g/mol
Cat. No.:
77694-621