/
Puerto Rico
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Chromoionophore VI, TRC, LGC Standards
77694-068
:
LGC Standards
New Product
:
Chromoionophore VI, TRC, LGC Standards
77694-068
:
LGC Standards
:
TRC-C432930-5MG
New Product
:
$204.55
Each(1/Itm/Bx)
-
+
CAS No.:
138833-47-3
Description:
Chromoionophore VI
Size:
5 mg
Formula:
C₃₈H₄₆Br₂O₅
Molecular Weight:
742.58 g/mol
Cat. No.:
77694-068