Emergency Burn Station, Certified Safety
Supplier: Certified Safety Manufacturing
216-033
56612-644CS 17.57 USD
56612-644
Emergency Burn Station, Certified Safety
Provides emergency burn dressings and ointments in a convenient heavy-gauge polystyrene display.
  • Complete with all the products needed for a burn emergency
The kit contains Certilizer™ eye and skin neutralizer for washing the area and neutralizing the effects of acid and alkali burns, Certicaine™ burn spray with aloe vera to alleviate minor pain associated with burns, and burn cream to prevent infection. Also included in the emergency burn station are assorted bandages, dressings, cold packs, a burn dressing kit, and an eye dressing kit.
Información para pedidos: Kit contents are pre-packaged in convenient replaceable units.