Order Entry
McPherson-Vannas Spring Scissors
New Product
McPherson-Vannas Spring Scissors
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  •  
    Length
    Material
    Blade length
    Tip size
  • McPherson-Vannas spring scissors, straight
    McPherson-Vannas spring scissors, straight
    77661-768
    14124
    8 cm (3")
    Stainless steel
    5 mm
    0.1 mm
    Each
    Each - $438.86
    $438.86
    Each
       $438.86
    New Product
    Specifications
    Environmentally Preferable
    New Product
  • McPherson-Vannas spring scissors, straight
    McPherson-Vannas spring scissors, straight
    77661-770
    14124-G
    8 cm (3")
    Premium German steel
    5 mm
    0.1 mm
    Each
    Each - $522.02
    $522.02
    Each
       $522.02
    New Product
    Specifications
    Environmentally Preferable
    New Product
  • McPherson-Vannas spring scissors, curved
    McPherson-Vannas spring scissors, curved
    77662-450
    501234
    8 cm (3")
    Stainless steel
    5 mm
    0.1 mm
    Each
    Each - $438.86
    $438.86
    Each
       $438.86
    New Product
    Specifications
    Environmentally Preferable
    New Product
  • McPherson-Vannas spring scissors, curved
    McPherson-Vannas spring scissors, curved
    77662-452
    501234-G
    8 cm (3")
    Premium German steel
    5 mm
    0.1 mm
    Each
    Each - $522.02
    $522.02
    Each
       $522.02
    New Product
    Specifications
    Environmentally Preferable
    New Product