Would you like to log in again?
VWR® Signature™ Diurnal Growth Chamber, Model 2015
Supplier: VWR International
VWR® Signature™
2015-2 2015
35960-078EA 8821.5 USD InStock
35960-078 35960-076
VWR® Signature™ Diurnal Growth Chamber, Model 2015
Incubators Cooled Incubators
The VWR® Signature™ Diurnal Growth Incubator is designed for studies requiring day and nighttime simulation.
  • Microprocessor temperature control
  • Dual program
  • Easy keypad set and calibration
  • CFC-free refrigerant
  • Diurnal illumination
  • Interior electrical outlet
  • VWR Two-Year Limited Parts and Labor Warranty
The unit is equipped with a hermetically-sealed compressor and an independent over temperature safety controller
This unit features dual-program selector dials, which allows control of two temperature conditions and an ON/OFF illumination cycle relative to the program selected. Each system operates independently allowing for simulation of a diurnal cycle, such as an eight hour day cycle of 30°C with light followed by a sixteen hour night cycle of 18°C without light. Forced air circulation ensures the most reproducible test conditions. The chamber air is gently and continuously circulated at a rate that ensures temperature uniformity (+/- 0.5°C at 20°C) of all test samples.
In addition, a one amp interior outlet allows use of shakers, stirrers, roller bottles or other apparatus. This chamber is ideal for plant growth studies.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
PRODUCTOS COMPLEMENTARIOS
SERVICIOS
Servicios: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty