Puerto Rico
Order Entry
Rat Recombinant IL-10 (Animal free) (from E. coli)
New Product
 
undefined
Rat Recombinant IL-10 (Animal free) (from E. coli)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Species:
    Rat
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    P29456
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Cytokine synthesis inhibitory factor|CSIF
  • UniProtKB:
    P29456
  • Protein/Peptide Name:
    IL-10 (AF)
  • Purity:
    ≥95%
  • Molecular Weight:
    18.8 kDa
  • Sequence:
    MSKGHSIRGD NNCTHFPVSQ THMLRELRAA FSQVKTFFQK KDQLDNILLT DSLLQDFKGY LGCQALSEMI KFYLVEVMPQ AENHGPEIKE HLNSLGEKLK TLWIQLRRCH RFLPCENKSK AVEQVKNDFN KLQDKGVYKA MNEFDIFINC IEAYVTLKMK N
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT

 

 

  •  
     
    Pack type
    Size
     
     
  • 77660-892
    300-35AF-100UG
    Vial
    100 µg
       
       
    Each
    Each - $802.37
    $802.37
    Each
       $802.37
     

     
    New Product
    New Product
  • 77660-894
    300-35AF-500UG
    Vial
    500 µg
       
       
    Each
    Each - $2,975.67
    $2,975.67
    Each
       $2,975.67
     

     
    New Product
    New Product