VWR® Fully Automatic Switchover Manifolds for Noncorrosive Gases
Supplier: VWR, part of Avantor
VWR®
4702665 4702658
82023-794EA 23979.87 USD
82023-794 82023-780
VWR® Fully Automatic Switchover Manifolds for Noncorrosive Gases
Gas Regulators
Manifolds keep a constant supply of gas to instrument to eliminate downtime.
  • Up to 225 psi delivery
  • Made of high-purity brass
A remote control panel will operate up to 30.5m (100') from the manifold. A remote alarm will operate up to 305m (1000') away from the manifold. LED indicators on the manifold box, control panel, and remote alarm display the active manifold bank and warn of empty banks. Remote alarm also features an audible alarm to indicate status of each bank.
Información para pedidos: Supplied with manifold box, control panel, and remote audio/visual alarm. Manifolds for one cylinder are supplied with two 61 cm (24") pigtails and headers; manifolds for two cylinders are supplied with four 61 cm (24") pigtails and headers.