VWR® Ground Joint Thermometers
Supplier: VWR, part of Avantor
VWR®
608041711 60170-4411 60804-1711 60170-4511
10186-032EA 97.18 USD
CA89095-786 10186-032 89095-786 10186-034
VWR® Ground Joint Thermometers
Thermometers Glass Thermometers Glass Thermometers with Standard Ground Joint
Calibrated for different depths of immersion, as measured from bottom of ground joint to bottom of bulb.
  • White back
  • Precise measurement
  • Celsius scale
With [ST]10/30 ground joint. Graduated in 1° divisions. Accuracy: ±1° to 150°C; ±1.5° from 150 to 250°C. Spirit-filled. Thermometer 89095-786 is fluoropolymer coated.