PYREX® Disposable Serological Shorty Pipets, Glass, Sterile, Plugged, Corning®
Supplier: Corning
PYREX®
7077B-25 7078B-25 7078B-50 7077B-50 7077B-1 7078B-1 7077B-10 7078B-5 7078B-10 7077B-5
53284-391CS 495.98 USD
53284-391 53284-449 53284-427 53284-463 89091-120 89091-268 53224-610 89091-224 89091-488 53284-460
PYREX® Disposable Serological Shorty Pipets, Glass, Sterile, Plugged, Corning®
Pipets Serological Pipets
These shorty glass pipets, calibrated 'to deliver' with blow-out, are ideal for use in tissue culture work and other procedures performed under a hood or in other confined areas
The 5, 10, and 25 ml sizes have tooled tops; the 50 ml size has a polypropylene top end.
Negative graduations make the pipet applicable for both serological and measuring procedures. Compatible with pipetting devices. ISO color coding is printed directly on the tear strip for easy identification.
Información para pedidos: Available individually wrapped in easy-open sterile wrappers or bag packed.