Would you like to log in again?
Clean-Ceil™ 2 Fan Filter Module, Microzone
Supplier: DESIGN FILTRATION MICROZONE CORP US
Clean-Ceil™
PFH-24 C2-24-H-A
35995-768EA 630.33 USD InStock
35995-768 35995-766
Clean-Ceil™ 2 Fan Filter Module, Microzone
Prefabricated Rooms
Designed to provide particle-free air for a variety of applications, including plenum or ducted cleanroom systems and portable cleanroom systems
When prefiltration of the air supply is necessary, the optional prefilter assembly (35995-768) is available. The assembly fits on top of the Clean-Ceil™ 2 module and draws air in through side intakes in the all-aluminum housing. These side intakes allow the Clean-Ceil™ 2 fan filter module to fit closely against the ceiling or other surface above the unit. The assembly contains two urethane foam prefilters that are treated with a fire retardant and rated to UL 94HF-1. They are washable, rated 40% efficient per ASHRAE 52-76, and attached with Velcro® fasteners for easy changing without the use of tools.
The integral motor/impeller assembly draws air from the surrounding area, forces it through a specially designed housing, and finally through a replaceable HEPA or ULPA filter. Standard module comes complete with solid-state speed control and electrical cord.
Información para pedidos: The Clean-Ceil™ 2 fan filter modules are also available with other combinations of filter types, filter replacement methods, and electrical requirements. Contact your VWR sales representative for details.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty