Profiller™ manual 435 Pipet Controller, DWK Life Sciences
Supplier: DWK Life Sciences
Profiller™
W835000
89044-770EA 270.8 USD
89044-770
Profiller™ manual 435 Pipet Controller, DWK Life Sciences
Pipets Pipet Controllers
Easy-to-use pipet controller features a lightweight design, ergonomic shape, and smooth control during aspirating or dispensing volumes of up to 100 ml with glass or plastic pipets.
Contoller also features a large-capacity egg-shaped bulb, soft-action lever button, and efficient blow-out system. Maintenance-free. Bulb color: gray.
Información para pedidos: Manufacturer's one-year warranty. For PTFE membrane filters, contact your Avantor representative.