Polyethylene glycol monomethyl ether, M.W. ∼500
Supplier: Thermo Scientific Chemicals
New Product
Certificates
About this item
:9004-74-4
:
:CH₃(OCH₂CH₂)nOH
:MFCD00084416
:12 °C
:1.08 g/cm³
:268 °C
:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:Colorless or white Liquid or viscous liquid or pasta
- Infrared spectrum:Conforms
- Water:≤1.0 %
- Average molecular weight:450 to 550
- pH:5.0 to 7.5 (1 % in water)
- Hydroxyl value:102 to 124 mg KOH/g
Specification Test Results