Puerto Rico
Order Entry
Ethyl-4-chloroquinoline-3-carboxylate ≥98.0% (by GC, titration analysis)
Ethyl-4-chloroquinoline-3-carboxylate ≥98.0% (by GC, titration analysis)
New Product
 
Ethyl-4-chloroquinoline-3-carboxylate ≥98.0% (by GC, titration analysis)
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    44.0 to 48.0 deg-C

 

 

  •  
     
    Size
     
     
  • TCE1201-200MG
    E1201-200MG
    200 mg
       
       
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $66.00
    $66.00
    Each(1/Itm/Bx)
       $66.00

     
    New Product
    New Product
  • TCE1201-1G
    E1201-1G
    1 g
       
       
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $196.00
    $196.00
    Each(1/Itm/Bx)
       $196.00

     
    New Product
    New Product