PharmaScoop® Scoops
Supplier: SAMPLING SYSTEMS LTD
New Product
Certificates
About this item
These fully welded, robust scoops are made from heavy gauge 316 stainless steel.
- Excellent corrosion resistance
- Heavy duty construction with satin finish
- Hygienic crevice free construction
- Radiused corners for easy cleaning
Please note: Length given in the table below is the body length (without handle).
Product Information
- Catalog #Supplier #CapacityLengthØAvailabilityPrice
- Specifications:New Product
Specifications
Environmentally PreferableNew Product - Specifications:New Product
Specifications
Environmentally PreferableNew Product - Specifications:New Product
Specifications
Environmentally PreferableNew Product - Specifications:New Product
Specifications
Environmentally PreferableNew Product - Specifications:New Product
Specifications
Environmentally PreferableNew Product - Specifications:New Product
Specifications
Environmentally PreferableNew Product - Specifications:New Product
Specifications
Environmentally PreferableNew Product - Specifications:New Product
Specifications
Environmentally PreferableNew Product - Specifications:New Product
Specifications
Environmentally PreferableNew Product - Specifications:New Product
Specifications
Environmentally PreferableNew Product - Specifications:New Product
Specifications
Environmentally PreferableNew Product