Falcon® 24-Well Multiwell Plates with Lids, Sterile, Corning®
Supplier: Corning
351147 351147
15705-060CS 217.51 USD
CA15705-060 15705-060
Falcon® 24-Well Multiwell Plates with Lids, Sterile, Corning®
Microplates Cell Culture Plates
This 24-well polystyrene plate is designed for assays not requiring cell attachment
The surface is nontreated for a hydrophobic growth surface and reduced cell attachment. Plate features flat-bottom wells with alphanumeric well identification, condensation rings, and a labyrinth lid for reduced evaporation and contamination. Sterilized by gamma irradiation and pyrogen-free.
Información para pedidos: Individually packaged in convenient peel-open, recyclable polyethylene terephthalate trays.