VWR® & VWR® Signature™ Extra Volume Pipette Tips
Supplier: VWR
Versapro
1168-960-306 1045-965-306 1168-800-000 1045-800-000
37001-204CS 730.56 USD
37001-204 37001-200 53508-920 53508-922
VWR® & VWR® Signature™ Extra Volume Pipette Tips
Pipette Tips
Designed to simplify aspirating from and dispensing into PCR tubes and plates.
  • VWR Signature™ tips and all sterile tips are endotoxin-free
  • Sterile tips are gamma-irradiated
  • Tips are RNase-, DNase-, and metal-free
  • Tips and hinged racks are autoclavable to 122 °C (252 °F)
Manufactured from polypropylene. Tips have ultrafine points and up to 250 µl of headspace to prevent overaspiration and pipettor contamination. Low-binding tips feature a modified polypropylene resin that limits the adhesion of cells, rDNA, and other viscous materials. Low-binding tips are also protease and human DNA-free.