VWR® & VWR® Signature™ Bevel Point Pipette Tips
Supplier: VWR
Versapro
1030-800-300 1039-800-000 1065-800-000 1030-900-300 1030-965-306 1060-800-000 1030-960-306
53508-794CS 993 USD
53508-794 53508-783 53509-122 53503-294 53508-810 53508-812 53503-566
VWR® & VWR® Signature™ Bevel Point Pipette Tips
Pipette Tips
VWR® and VWR® Signature™ Bevel Point pipette Tips provide maximum sample recovery and minimal fluid retention.
  • For use with most pipettors
  • Sterile tips are endotoxin-free
  • Available in sterile and non sterile formats
  • All tips are free of RNase, DNase, and trace metals
They have a beveled end for superior accuracy and reproducibility in microliter volume ranges. Low-binding tips feature a modified polypropylene resin that limits the adhesion of cells, rDNA, and other viscous materials. Low-binding tips are also free of protease and human DNA.
Tips and hinged racks are fully autoclavable to 122 °C (252 °F).
Información para pedidos: Pak Rak® Express refills contain pipette tips to fill 8×12 racks, without the packaging waste of standard tip boxes. Pak Rak Express refills require the Pak Rak system loader (see 30128-374 series), which dispenses tips quickly and easily from the refill pack into each rack, eliminating risk of contamination from hands or gloves.