1536-Well Microplates, Corning
Supplier: Corning
3726BC 3725 3833 3831 3835 3895 3832BC 3893BC 3937 7246 7249 3838 3726 3836 3838BC 3936 3729BC 3728BC 3724 3725BC 3728 3839BC 7248 3724BC 3727BC 3891 3893 3731BC 3729 3727 7247
89090-260CS 1642.99 USD
89090-260 89218-982 89218-980 89423-138 89089-728 89089-738 89130-734 89089-734 66025-652 89089-732 71001-346 66025-654 89130-730 89089-730 89218-978 89126-916 71001-348 89126-918 89089-726 71001-350 89089-748 71001-352 89089-724 89089-746 77776-916 30621-094 89089-744 89089-742 89130-726 89089-740 77776-918
1536-Well Microplates, Corning
Microplates HTS Microplates
These polystyrene microplates offer four times the density as 384-well plates and are ideal for use in high throughput screening procedures
Microplates offer a total working volume of 10µL for round well bottom vessels, and 12.8µL for flat well bottom vessels. Round well bottom microplates are designed to reduce air entrapment and wicking, while offering improved CV and Z factors.
Plates can be used for a wide array of HTS miniaturized applications, such as general fluorescence assays, fluorescence polarization, fluorescence polarization immuno assays, time resolved fluorescence, fluorescence resonance energy transfer, luminescence assays, and time resolved luminescence assays. Microplates conform to industry standard footprint and dimensions, and are available in solid black or white with round or flat bottoms in black or clear bottom formats. Available surface treatments include tissue culture treated, nonbinding (NBS®) and untreated.