Falcon® Multiwell Flat-Bottom Plates with Lids, Sterile, Corning®
Supplier: Corning
353075 353043 353226 353935 353046 353225 353072 353934 353936 353224 353224 353936 353046 353075 353047 353047 353230 353935 353072 353078 353934 353226 353078 353230 353225 353043
62406-165CS 183.85 USD
CA62406-155 CA62406-157 CA62406-195 62406-165 25382-204 CA62406-117 CA25382-198 21100-006 21100-004 CA62406-081 CA62406-161 CA62406-183 62406-195 21100-002 62406-159 CA62406-165 62406-155 CA25382-204 62406-117 CA21100-004 CA21100-002 25382-198 CA21100-006 62406-161 62406-183 62406-081
Falcon® Multiwell Flat-Bottom Plates with Lids, Sterile, Corning®
Microplates Cell Culture Plates
These polystyrene plates have a consistent tissue culture surface chemistry produced by using vacuum-gas plasma treatment
Each plate features a patented low evaporation lid, deep wells, serrated gripping panels, a large writing patch, and alphanumeric well identification. Sterilized by gamma irradiation.
Exceptional optical clarity allows for visualization of cell monolayers. They are designed for manual and automated handling. Plates can be stacked together for efficient use of incubator space.
Información para pedidos: Plates with lids are packaged in peel-apart packaging to assure sterile delivery. Plates 21100-002, 21100-004, and 21100-006 feature a ready-stack bulk tray, which is designed to handle a large number of plates aseptically.