96-Well Half-Area Plates, Polystyrene, Corning®
Supplier: Corning
3697 3688 3642 3994 3885 3884 3880 3693 3993 3886 3992 3883 3695 3881 3686 3690 3694 3875 3696 3882
33501-002CS 1146.29 USD
33501-002 29444-320 29444-314 33501-004 29444-312 33501-006 29444-308 29444-316 33500-998 29442-318 33501-818 77776-924 33501-808 33500-996 33500-994 33501-008 33501-834 33501-814 29442-062 33501-816
96-Well Half-Area Plates, Polystyrene, Corning®
Microplates Cell Culture Plates
These flat-bottom plates have well centers in the same position as standard 96-well plates for use in existing automated systems, including plate readers and washers, and pipetting equipment
Tissue culture plates are treated to facilitate uniform cell attachment for cell-based assays.
White plates have low background luminescence. Black plates have low background fluorescence, enabling readings down to 340nm. Nonbinding surface (NBS®) plates are ideal for reducing protein and nucleic acid binding at low concentrations and increasing assay signal-to-noise.
Sterile plates are sterilized by gamma radiation and certified nonpyrogenic.
Increased distance between the wells minimizes well-to-well crosstalk.