TF (PTFE) Membrane Disc Filters, Cytiva (Formerly Pall Lab)
: Avantor
Excellent for air and gas filtration, and for venting applications.
- Chemically resistant
- HPLC certified
- Hydrophobic
HPLC certified for use in nonaqueous mobile phase filtration and degassing. Hydrophobic PTFE on a polypropylene support. Filter 28150-804 includes 37 mm support pads.
Highly resistant to chemical attack.