Culture dishes for in vitro fertilisation, Falcon®
Supplier: Corning
Falcon®
353653 353655
25382-194CS 2637.42 USD
25382-194 89511-923
Culture dishes for in vitro fertilisation, Falcon®
Culture Dishes Petri Dishes
PS, sterile, with lid. Pre-tested and certified plasticware for in vitro fertilisation (IVF), designed for consistency and ease of use. These dishes have perfectly flat, optically clear surfaces for optimum manipulation and observation of ova and embryos. Lids are designed for aseptic manipulation and consistent venting to maintain humidification.
  • Non-embryotoxic, non-pyrogenic and non-cytotoxic
  • Tissue culture treated for a consistent hydrophilic surface
  • Sterilised by gamma irradiation
  • Packaged in peel-open, medical-style packaging
  • Multi-unit bags have reseal tabs
They have flat, optically clear surfaces for optimum manipulation and observation. Lids are designed for aseptic manipulation and consistent venting to maintain humidification. Sterilized by gamma irradiation. Nonembryotoxic (the lids of the Falcon IVF Products are not certified nonembryotoxic), nonpyrogenic, and noncytotoxic.
Información para pedidos: Dishes are supplied with lids and packaged in resealable bags.