Culture dishes for in vitro fertilisation, Falcon®
Supplier: Corning
Falcon®
353652 353652
25382-192CS 843.46 USD
25382-192 CABD353652
Culture dishes for in vitro fertilisation, Falcon®
Culture Dishes Cell Culture Dishes
PS, sterile, with lid. Pre-tested and certified plasticware for in vitro fertilisation (IVF), designed for consistency and ease of use. These dishes have perfectly flat, optically clear surfaces for optimum manipulation and observation of ova and embryos. Lids are designed for aseptic manipulation and consistent venting to maintain humidification.
  • Non-embryotoxic, non-pyrogenic and non-cytotoxic
  • Tissue culture treated for a consistent hydrophilic surface
  • Sterilised by gamma irradiation
  • Packaged in peel-open, medical-style packaging
  • Multi-unit bags have reseal tabs
Dishes are nonpyrogenic, noncytotoxic, nonembryotoxic, and sterilized via gamma irradiation.
Información para pedidos: Dishes are supplied with lids and packaged in resealable bags.