VWR® Scintillation Vials, PE, with Screw Cap
Supplier: Avantor
VWR®
VW58515-20 VW58504-20 VW58501-20 58510B-20
66022-241CS 395.76 USD
66022-241 66022-274 66022-263 66022-222
VWR® Scintillation Vials, PE, with Screw Cap
Vials
Polyethylene meets rigid specifications for low background counts, resulting in a container especially suitable for low activity counting.
  • Polyethylene construction
  • Fit all standard liquid scintillation counting equipment
  • Choose attached or unattached caps
Each vial includes a white screw cap with a top suitable for marking.
Información para pedidos: Tray packed vials are packed in 100-place cellular trays, which have alphanumeric coordinates for easy sample identification.