Thermowell® Individual PCR Tubes, Corning®
Supplier: Corning Life Sciences
Thermowell®
6530 6531 6571
53495-756CS 147.75 USD
53495-756 29443-340 29442-340
Thermowell® Individual PCR Tubes, Corning®
Tubes PCR Tubes
Polypropylene tubes feature a thin wall and are designed for precise fit in heat blocks, to ensure fast, uniform heat transfer.
  • Autoclavable at 121 °C (250 °F)
  • Certified DNase- and RNase-free
  • Natural color
The 0.6 ml tubes are compatible with all 0.5 ml heat blocks. The 0.2 ml tubes are compatible with all 0.2 ml heat blocks.