SP Bel-Art SMORC® Small Orifice Cleaner, Bel-Art Products, a part of SP
Supplier: Bel-Art
SMORC®
F38126-0000
60380-941PK 90.27 USD
60380-941
SP Bel-Art SMORC® Small Orifice Cleaner, Bel-Art Products, a part of SP
Sprays
Ideal for cleaning recorder pens, syringe needles, burets, valves, micropipettors, and other small orifices.
  • Effectively cleans small orifices
  • Ring magnet in handle picks up wires
  • Secure stainless steel wire for reliable cleaning
A pin vise holds the stainless steel wire securely while cleaning. A ring magnet on the handle is useful for picking up wires.
Handle length: 102 mm (4")
Información para pedidos: Cleaner is supplied with a package of assorted stainless steel wires, 10 each of three diameter sizes: 0.152, 0.254, and 0.508 mm (0.006, 0.010, and 0.020"). Replacement wires are supplied in a plastic vial containing the same size assortment.