Would you like to log in again?
Adjustable Thermopolastic Wrist Strap, SCS
Supplier: 3M ELECTRONIC SPECIALTY MARKET
051138-55697
12778-060EA 21.79 USD InStock
12778-060
Adjustable Thermopolastic Wrist Strap, SCS
Grounders Wrist Grounders
The 4600 Wristband Series consists of an insulative exterior with conductive interior and 4 mm metal male snap. Made of thermoplastic material, the wristband is lightweight, low profile, and makes contact with human skin, assuring employee acceptance while helping to ensure ease of use.
  • >1×107 ohm exterior with conductive interior <1×105 ohms
  • 5' coil cord, with one megohm current limiting resistor located at socket end
  • Assembled in the United States of America
The 'zipper' style latching mechanism easily adjusts to most wrist sizes.
Certificaciones: Meets requirements of ANSI/ESD S20.20 and Wrist Strap standard ANSI/ESD S1.1. Meets EN 61340-5-1 and IEC 61340-4-6.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty