CONSTIX®, Sealed Polyester Swabs, SP-5 Series, Contec®
Supplier: Contec
CONSTIX®
SP-5
89107-992CS 105.15 USD
89107-992
CONSTIX®, Sealed Polyester Swabs, SP-5 Series, Contec®
Swabs Swabs for Cleaning
The CONTIX® SP-5 is a sealed polyester swab consisting of 100% knit polyester fabric thermally bonded to a polypropylene handle.
  • Polyester head features extremely low particulate and residue levels
  • Polypropylene handle provides excellent resistance to most common solvents, acids, and bases
  • Flexible tip provides a softer application on surfaces
The head of the swab is thermally attached to the handle without adhesives or binders, and is made from cleanroom laundered polyester fabric. The SP-5 swab is durable, absorbent, and features extremely low particulate and residue levels.