Matrix® No-Roll Earplugs, Honeywell Safety
Supplier: Honeywell Safety
Matric®
MTX-30-OR MTX-1-OR-D MTX-1-OR
CA82020-870 CA82020-880 CA82020-886
Matrix® No-Roll Earplugs, Honeywell Safety
Earplugs Disposable Earplugs
Disposable earplugs are constructed of a dual-density foam with a soft outer skin that is gentle on the sensitive skin inside the ear canal
Matrix earplugs have a smaller diameter than other earplugs, meaning no rolling is necessary for a fast and easy insertion. There is no need to wait for the foam to expand; earplugs deliver instant protection upon proper insertion. A uniform attenuation profile blocks loud noise while allowing voice and other high freqencies to be heard more naturally. Three attenuation levels allow targeted attenuation.