Stereo Zoom Microscope, SMZ Series, Motic®
Supplier: Motic
1101000900091 1101001400673 1101000601161 1101001400673 1101000900071 1101000900091 1101000601161 1101000900071 1101000200252
19000-498EA 101.85 USD
CA19000-418 19000-498 19000-080 19000-414 CA19000-416 19000-416 CA19000-414 CA19000-080 19000-418
Stereo Zoom Microscope, SMZ Series, Motic®
Microscopes
The stereo zoom microscope has bilateral zoom control knobs for comfort and flexibility of use.
Microscope features rugged, die-cast metal construction, diopter adjustment on both eyetubes with interpupillary distance adjustment between 54 and 76mm, and a pair of widefield highpoint 10x, 20mm eyepieces.Strong incident and transmitted halogen illumination is provided by a 12V, 10W bulb with intensity control.
The working distance is 80mm without auxiliary objective, and the continuous zoom range of 1x to 4x requires no refocussing through the entire magnification range.The SMZ-140 features a 45° inclined, 360° rotational binocular head. Industrial holder 19000-418 is for use with bonding machines.
Información para pedidos: Heads, focusing mounts, eyepieces, objectives, stands and other accessories sold separately.