DECON-PHENE® II, High pH Phenolic, Veltek Associates
Supplier: Veltek
DECON-PHENE®
DP2-07-16Z-02 DP2-02 DP2-03-2Z DP2-01 DP2-05-1Z DP2-06-16Z-02 DP2-07-16Z-01 DP2-03-4Z DP2-06-16Z-01 DP2-04-2Z DP2-04-1Z
14222-698CS 312.73 USD
14222-698 14003-668 75795-396 14003-666 14004-484 75795-618 14003-664 14004-486 14003-662 14003-660 14004-482
DECON-PHENE® II, High pH Phenolic, Veltek Associates
Cleaning Agents Detergents and Surface Cleansers
DECON-PHENE is a high pH, sodium based, phosphate-free, phenolic, synthetic germicidal detergent that is used where a broad spectrum disinfectant is required in moderate amounts of organic soil.
  • Filtered at 0.2 Microns
  • Double-bagged packaged
  • Gamma irradiated
Strong enough to kill a broad spectrum of pathogenic and non-pathogenic bacteria. Mild enough to have no harmful effect on the surface being disinfected. Delivered with lot specific Certificate of Analysis, and Sterility Report, Certificate of Irradiation.