Would you like to log in again?
Cube-Shaped UN Rated Poly Drum, New Pig
Supplier: NEW PIG CORP
DRM796 DRM794
56620-736EA 121.59 USD InStock
56620-736 56620-732
Cube-Shaped UN Rated Poly Drum, New Pig
Kegs
  • Heavy-duty, open-head drum is approved for domestic and international shipments
  • Made of reusable/recyclable high-density polyethylene Molded-in handles make maneuvering quick and easy
  • FDA-approved for shipment and containment of food products
  • Color: blue with black lid
Cubic design makes shipping and storing easier than traditional round drums. Heavy-duty poly construction is reusable and recyclable. FDA Approved resins for containment of food products.
Locking metal ring and polyethylene lid with rubber gasket seal drum tight. Molded-in handles allow easy maneuvering.
Certificaciones: UN Certification 3H2/Y90/S is approved for domestic and international shipments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty