A cost-effective, highly efficient auto-fill liquid nitrogen (LN2) tank.
- Auto-fill convenience
- Extremely low LN2 consumption
- Remote alarm contacts
- Vapor storage temperature of –185 °C (–301 °F) or colder
- VWR two-year limited parts and labor and five-year limited vacuum warranty
Unique reduced opening neck design, hanging racks, and high vacuum insulation provide extremely low vapor temperatures of –185 °C (–301 °F) or colder using only a few inches of LN2
CryoRetriever claw-style tool is specifically designed for retrieving boxes, vials, and frames from cryopreservation tanks. Exclusive sealed center shaft prevents hazardous spurting of LN2 that can occur with a hollow shaft retrieval claw.
Typically requires less than one LN2 supply cylinder per month. Easy-to-use controller features digital temperature display, audible and visual low level alarms, low supply and high tank temperature alarms and remote alarm contacts. Unit is mounted on 5 wheeled caster platform and includes a locking provision. Under factor setting, 90% of sample capacity is in vapor phase storage.
Información para pedidos: Cryostorage unit only includes seven stainless steel hanging racks. Each rack has a capacity of 10 boxes (not included). Package systems include cryostorage unit, full inventory system (stainless steel hanging racks, 100-cell boxes, and box dividers), 1.8 m (6') stainless steel transfer hose, liquid level measuring stick, and a pair of cryo-gloves. Professional installation and setup is available and highly recommended for this product. Add this item to your order to receive on-site initial start-up, controller settings and an employee inservice by a factory representative.
Precaución: Note: Static Evaporation Rates and Static Holding Times are nominal. Actual rates may be affected by the nature of the contents, atmospheric conditions, container history, and manufacturing tolerances.