Corning® Phenolic Screw Caps, Disposable, Rubber-Lined
Supplier: Corning Life Sciences
Corning®
99999-18 99999-15
60826-494CS 294.02 USD
60826-494 60826-483
Corning® Phenolic Screw Caps, Disposable, Rubber-Lined
Closures Test/Sample Tube Closures
These phenolic screw caps have a white rubber lining and are designed to fit PYREX® screw-top tubes with a matching cap size (reusable or disposable)
Autoclavable.