Airstream® Headgear-Mounted Powered Air Purifying Respirator, 3M™
AS-400LBC AS-400LBC
56221-876PK 2433.37 USD
56221-876
Airstream® Headgear-Mounted Powered Air Purifying Respirator, 3M™
Powered air purifying respirator (PAPR) is lightweight and comfortable and requires no breathing tubes or hoses.
The smart battery charger can fully charge up to five discharged batteries in approximately eight hours. It features LED status of each battery and automatic switchover to trickle rate mode to prevent overcharging.
Caution: All 3M products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
Positive pressure helps keep out particles and keep the face shield 'fog-free.' The blower and filter are mounted inside the headgear shell and powered by a rechargeable nickel cadium battery pack, which clips to the user’s belt. A head seal, faceseal, and temple seals help prevent mixing of filtered air inside the visor with ambient air. When not in the contaminated area, the visor assembly can be lifted and locked in the up position. Respirator will accommodate limited facial hair without compromising the level of protection. Respirator helps provide protection against airborne particulate contaminants. NIOSH-approved.