About this item
Eppendorf rotors for centrifuge 5430/5430 R, precision at its core. Maximize your centrifuge's potential with rotors crafted for efficiency. Tailor your experiments with various capacities and speeds, ensuring optimal sample processing.
- Precision-engineered for optimal sample separation
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 minutes)
- Accessories (adaptors and buckets) further customize your centrifuge setup
- Aerosol-tight and PTFE coated options available
Explore enhanced centrifugation possibilities with Eppendorf rotors designed for centrifuge 5430/5430 R. Choose from a range of capacities and speeds, providing versatility for microcentrifugation or larger sample volumes. The precision-engineered rotors ensure optimal sample separation. Accessories such as adaptors and buckets further customize your centrifuge setup. Maximize efficiency with the ability to handle diverse sample types. Elevate your laboratory's capabilities, achieve reproducible results, and enjoy the flexibility to meet specific application needs.
Specifications
- Catalog No:
- 80094-204
- 80094-188
- 80094-208
- 89423-576
- 80094-196
- 89423-578
- 80094-200
- 80094-210
- 80094-184
- 89423-580
- 80094-192
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental Certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001