DuPont™ ProClean® Hoods with Full Face Opening
PC663SWH000B PC663SWH000B
13452-580CS 227.62 USD
13452-580 CA13452-580
DuPont™ ProClean® Hoods with Full Face Opening
ProClean® offers improved non-hazardous light liquid splash protection.
  • Serged seams
  • Full face opening
  • Bound hood opening with ties
  • One size fits most
ProClean® is made from a microporous composite fabric. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Bound hood opening ensures lower particle shedding. Full face, 10 inch opening with bound ties allows one size to fit most.
Envase: Bulk packaged garments are bulk packaged in double transparent poly liners.