WaterPro® PS Polishing Stations, Labconco®
Supplier: Labconco
WaterPro®
9047101
30181-416EA 1067.7 USD
30181-416
WaterPro® PS Polishing Stations, Labconco®
Water Purification Systems Deionizers
Designed for general chemistry labs requiring Type I (up to 18.2 megohm-cm) water
Models 26301-853 and -672 have a dispensing valve. Models 26301-860 and -674 have the dispensing valve and hand-held dispensing gun with 0.91m (3') recirculating line. Dispensing module(s) are incorporated within the polishing station — no separate module is required.
Quiet system with 1/4hp pump and in-line pressure regulator has large pure polypropylene bowls for one activated carbon and three deionization cartridges (not included). Automatic continuous recirculation through the polishing loop maintains water quality, minimizes bacterial growth, and reduces rinse up time. Polishing station includes a power switch, LED water purity level display, and a timed control dispensing valve which offers delivery up to 1.8L per minute. Cabinet measures 79.1Wx19.6Dx21.9Hcm (311/8x73/4x85/8"). The overall height with bowls attached is 73.3cm (287/8"). Polishing station requires polishing kit 30181-416, which includes a carbon cartridge and three deionization cartridges.
Certificaciones: The 115V models are ETL and ETL-C listed. The 230V models are CE marked.
Información para pedidos: Manufacturer's one-year warranty on parts and labor. Choose 115V with 2.4m (8') three-wire cord and plug, or 220V with 2.4m (8') three-wire cord (plug required). Optional hollow fiber final filter can be used on dispensing valve or gun to remove all particulates and bacteria larger than 0.2µm in diameter.