PYREX® Lifetime Red™ Graduated Cylinders, To Contain, with Reinforced Glass Bead, Corning
Supplier: Corning
PYREX® Lifetime Red™
3046-250 3046-25 3046-10 3046-100 3046-50
89090-926CS 977.34 USD
89090-926 89090-832 89090-634 89090-802 89090-622
PYREX® Lifetime Red™ Graduated Cylinders, To Contain, with Reinforced Glass Bead, Corning
Hexagonal base prevents rolling
The 100mL size is in accordance with ASTM E133 and E1272, for D86, D216, and D447.
A reinforced bead of glass near the top helps to reduce breakage if the cylinder upsets. Fire polished rim, easily read white graduations enameled against a red panel. Single metric scale.