Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (TRITC)
- Catalog No:
- 77022-532
- 77021-672
- Antigen Symbol:
- CD3e
- CD3e
- Antigen Name:
- CD3e antigen
- CD3e antigen
- Antibody Type:
- Secondary
- Secondary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- TRITC (Tetramethylrhodamine Isothiocyanate)
- TRITC (Tetramethylrhodamine Isothiocyanate)
- Clone:
- SK7
- SK7
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Purification:
- Affinity purification
- Affinity purification
- Storage Temperature:
- Refrigerated (2...8 °C); Minimize light exposure
- Refrigerated (2...8 °C); Minimize light exposure
- Shipping Temperature:
- 20
- 20
- SizePackaging
- Minority or Woman-Owned Business EnterpriseSmall Business Enterprise
Specifications
Antigen SymbolCD3eAntigen NameCD3e antigenEnvironmentally PreferableAntibody TypeSecondaryClonalityMonoclonalConjugationTRITC (Tetramethylrhodamine Isothiocyanate)CloneSK7ReactivityHumanHostMouseIsotypePurificationAffinity purificationStorage TemperatureRefrigerated (2...8 °C); Minimize light exposureShipping Temperature20Minority or Woman-Owned Business EnterpriseSmall Business Enterprise - Minority or Woman-Owned Business EnterpriseSmall Business Enterprise
Specifications
Antigen SymbolCD3eAntigen NameCD3e antigenEnvironmentally PreferableAntibody TypeSecondaryClonalityMonoclonalConjugationTRITC (Tetramethylrhodamine Isothiocyanate)CloneSK7ReactivityHumanHostMouseIsotypePurificationAffinity purificationStorage TemperatureRefrigerated (2...8 °C); Minimize light exposureShipping Temperature20Minority or Woman-Owned Business EnterpriseSmall Business Enterprise