Erlenmeyer Flasks with Screw Caps, PMP, BrandTech
Supplier: BrandTech
V66895 V66795
83008-944PK 136.89 USD
83008-944 83008-948
Erlenmeyer Flasks with Screw Caps, PMP, BrandTech
Flasks Erlenmeyer Flasks
Flasks feature a wide-mouth design for easy pouring and a printed scale of approximate volumes.
  • Break-resistant and easy-to-clean
  • Autoclavable
  • Compatible with ground joint stoppers (not included)
Made of transparent PMP (polymethylpentane). Supplied with a polypropylene screw cap.