Ideal for both QC and research applications in lab, plant, or field.
- Extra-tough, shockproof ABS plastic construction
- Multiple timer features
- Traceable®
Stopwatch times to 9 hours, 59 minutes, 59 seconds, 99 hundredths.
Unique feature allows repeat countdown alarm timing. Set it to countdown at any time from 10 hours to 0.1 second. When zero is reached, the timer alarms and again begins counting down from the programmed time. This repeat cycle continues automatically until stopped. LCD continually shows running time remaining to zero.
Provides stopwatch functions of single action, time-in/time-out, cumulative split, interval split and continuous timing.
Certificaciones: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Información para pedidos: Supplied with lanyard, instructions, and two-year, 1.5 V silver-oxide battery.