Order Entry
Idex® PEEK Tubings, 5 Ft. Length
undefined
Idex® PEEK Tubings, 5 Ft. Length
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  •  
    Ø int.
    Material
    Ext.Ø
    Color
    Length
    Max. pressure
  • Chromatography tubing
    Chromatography tubing
    100461-842
    02006-12
    0.001"
    PEEK
    0.014"
    Natural
    5 ft.
    345 bar (5000 psi)
    Each
    Each - $88.92
    $88.92
    Each
       $88.92
    Specifications
    Environmentally Preferable
  • Chromatography tubing
    Chromatography tubing
    100449-722
    02006-08
    0.004"
    PEEK
    ¹/₁₆"
    Black
    5 ft.
    483 bar (7000 psi)
    Each
    Each - $72.62
    $72.62
    Each
       $72.62
    Specifications
    Environmentally Preferable
  • Chromatography tubing
    Chromatography tubing
    100461-872
    02006-16
    0.005"
    PEEK
    ¹/₃₂"
    Red
    5 ft.
    345 bar (5000 psi)
    Each
    Each - $87.08
    $87.08
    Each
       $87.08
    Specifications
    Environmentally Preferable
  • Chromatography tubing
    Chromatography tubing
    100461-874
    02006-17
    0.007"
    PEEK
    ¹/₃₂"
    Yellow
    5 ft.
    345 bar (5000 psi)
    Each
    Each - $87.08
    $87.08
    Each
       $87.08
    Specifications
    Environmentally Preferable
  • Chromatography tubing
    Chromatography tubing
    100461-882
    02006-18
    0.010"
    PEEK
    ¹/₃₂"
    Blue
    5 ft.
    345 bar (5000 psi)
    Each
    Each - $87.08
    $87.08
    Each
       $87.08
    Specifications
    Environmentally Preferable
  • Chromatography tubing
    Chromatography tubing
    100461-898
    02006-11
    0.020"
    PEEK
    ¹/₃₂"
    Orange
    5 ft.
    345 bar (5000 psi)
    Each
    Each - $87.08
    $87.08
    Each
       $87.08
    Specifications
    Environmentally Preferable