Puerto Rico
Order Entry
 
6-Chloro-N-(3,4-dichlorophenyl)pyrimidin-4-amine 97%
  77248-216
 :  Ambeed
 :  
 
6-Chloro-N-(3,4-dichlorophenyl)pyrimidin-4-amine 97%
Catalog # 77248-216
Supplier:  Ambeed
Supplier Number:  A179182-1G
CAS Number:  
Price
$65.00
Log in for availability
Unit of Measure Each (1g)
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 g
  • Cat. No.:
    77248-216

Specifications

 

 :
 :274.53 g/mol
 :GHS07
 :6303-46-4
 :C₁₀H₆Cl₃N₃
 :MFCD14611159